Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myb
Entrez Gene ID
17863
Gene Name
myeloblastosis oncogene
Gene Aliases
AI550390, M16449, c-myb
Chromosome Location
Chr.10: 21124930 - 21160984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198914.1 NP_001185843.1 6 - 7 1033 61
NM_010848.3 NP_034978.3 6 - 7 1033 61
GenBank mRNA AK088079.1 - 6 - 7 954 61
AK088598.1 - 6 - 7 955 61
AK132250.1 - 6 - 7 898 61
AK137965.1 - 6 - 7 899 61
BC011513.1 - 6 - 7 941 61
X02774.1 - 6 - 7 803 61

View Full Product Details