The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110129.1 | NP_001103599.1 | 5 - 6 | 289 | 100 |
NM_001110130.1 | NP_001103600.1 | 5 - 6 | 289 | 100 | |
NM_028677.4 | NP_082953.1 | 5 - 6 | 289 | 100 | |
GenBank mRNA | AK003179.1 | - | 5 - 6 | 267 | 100 |
AK005202.1 | - | 5 - 6 | 250 | 100 | |
AK008394.1 | - | 5 - 6 | 267 | 100 | |
AK014665.1 | - | 5 - 6 | 250 | 100 | |
AK134995.1 | - | 5 - 6 | 269 | 100 | |
AK136330.1 | - | 5 - 6 | 248 | 100 | |
AK164299.1 | - | 5 - 6 | 254 | 100 | |
BC016565.1 | - | 5 - 6 | 244 | 100 | |
BC050116.1 | - | 5 - 6 | 243 | 100 | |
BC098490.1 | - | - | 245 | 100 | |
BY708305.1 | - | 5 - 6 | 266 | 100 |