Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zscan20
Entrez Gene ID
269585
Gene Name
zinc finger and SCAN domains 20
Gene Aliases
C130001F22, Zfp31, Zkscan20
Chromosome Location
Chr.4: 128578670 - 128610172 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177758.4 NP_808426.2 2 - 3 611 78
GenBank mRNA AK047820.1 - 2 - 3 532 78
AK144249.1 - 2 - 3 612 78
AK153688.1 - 2 - 3 548 78
AK153845.1 - 2 - 3 548 78
BC076602.1 - 2 - 3 527 78

View Full Product Details