The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177077.2 | NP_796051.2 | 6 - 7 | 775 | 82 |
GenBank mRNA | AK090104.1 | - | 5 - 6 | 599 | 82 |
AK144547.1 | - | 6 - 7 | 776 | 82 | |
AK165019.1 | - | 6 - 7 | 720 | 82 | |
BC144781.1 | - | 6 - 7 | 718 | 82 | |
BC145693.1 | - | 6 - 7 | 718 | 82 |