Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Setmar
Entrez Gene ID
74729
Gene Name
SET domain without mariner transposase fusion
Gene Aliases
5830404F24Rik, Etet2
Chromosome Location
Chr.6: 108065045 - 108077127 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276356.1 NP_001263285.1 1 - 2 198 108

View Full Product Details