Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nsg2
Entrez Gene ID
18197
Gene Name
neuron specific gene family member 2
Gene Aliases
-
Chromosome Location
Chr.11: 32000386 - 32059211 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290680.1 NP_001277609.1 1 - 2 43 71
GenBank mRNA BU058771.1 - 1 - 2 43 71

View Full Product Details