The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028604.2 | NP_082880.2 | 1 - 2 | 156 | 62 |
GenBank mRNA | AK014181.1 | - | 1 - 2 | 86 | 62 |
AK032943.1 | - | 1 - 2 | 103 | 62 | |
BC048703.1 | - | 1 - 2 | 72 | 62 | |
BY259542.1 | - | 1 - 2 | 104 | 62 |