Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klrk1
Entrez Gene ID
27007
Gene Name
killer cell lectin-like receptor subfamily K, member 1
Gene Aliases
D6H12S2489E, NKG2-D, Nkg2d
Chromosome Location
Chr.6: 129610323 - 129623864 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083322.2 NP_001076791.1 1 - 2 56 80
NM_001286018.1 NP_001272947.1 1 - 2 56 80
GenBank mRNA AK170499.1 - 1 - 2 57 80

View Full Product Details