Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BC048546
Entrez Gene ID
232400
Gene Name
cDNA sequence BC048546
Gene Aliases
Ovos
Chromosome Location
Chr.6: 128539822 - 128581606 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001179.3 NP_001001179.2 23 - 24 2941 90
GenBank mRNA AK138836.1 - 23 - 24 2942 90
AK161068.1 - 10 - 11 1291 90
AK161184.1 - 23 - 24 2945 90
BC040781.1 - 11 - 12 1393 90

View Full Product Details