The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028995.3 | NP_083271.1 | 6 - 7 | 872 | 62 |
GenBank mRNA | AK018640.1 | - | 6 - 7 | 849 | 62 |
AK049696.1 | - | 6 - 7 | 907 | 62 | |
AK087811.1 | - | 6 - 7 | 875 | 62 | |
AK138843.1 | - | 6 - 7 | 873 | 62 | |
AK148206.1 | - | 6 - 7 | 893 | 62 | |
BC057168.1 | - | 6 - 7 | 812 | 62 |