Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
4831410D14
Entrez Gene ID
327901
Gene Name
uncharacterized protein 4831410D14
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK029182.1 - 1 - 2 239 66


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB613702.1 - 1 - 2 236 Chr. 11 - 33204057 - 33219366 [+] on Build GRCm38 66


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream RAN binding protein 17 Ranbp17 Anti-sense Chr. - 33211794 - 33514750 on Build GRCm38
Downstream acidic (leucine-rich) nuclear phosphoprotein 32 family, pseudogene Anp32-ps Anp32c, Anp32c-ps, Anp32cl, EG448829 178.9kb Anti-sense Chr. - 33394905 - 33395397 on Build GRCm38

View Full Product Details