The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277121.1 | NP_001264050.1 | 1 - 2 | 86 | |
GenBank mRNA | AK144682.1 | - | 1 - 2 | 101 | 86 |
AK152039.1 | - | 1 - 2 | 86 | ||
BC026850.1 | - | 1 - 2 | 84 | 86 |