Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1d
Entrez Gene ID
57316
Gene Name
C1D nuclear receptor co-repressor
Gene Aliases
1110036E10Rik, AI875855, SUN-CoR
Chromosome Location
Chr.11: 17257618 - 17269176 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020558.3 NP_065583.2 2 - 3 188 78
GenBank mRNA AK028702.1 - 2 - 3 195 78
AK035169.1 - 2 - 3 189 78
BC005436.1 - 2 - 3 184 78
X95591.1 - 2 - 3 215 78

View Full Product Details