Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pex7
Entrez Gene ID
18634
Gene Name
peroxisomal biogenesis factor 7
Gene Aliases
MmPEX7
Chromosome Location
Chr.10: 19860090 - 19907674 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161825.1 NP_001155297.1 4 - 5 535 66
NM_008822.2 NP_032848.1 5 - 6 613 66
GenBank mRNA AK004687.1 - 5 - 6 598 66
AK160195.1 - 5 - 6 577 66
BC132213.1 - 5 - 6 586 66
BC132503.1 - 5 - 6 586 66
BC145300.1 - 4 - 5 508 66
U69171.1 - 5 - 6 604 66

View Full Product Details