The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008229.2 | NP_032255.2 | 1 - 2 | 263 | 71 |
GenBank mRNA | AB221663.1 | - | 1 - 2 | 55 | 71 |
AK051753.1 | - | 1 - 2 | 238 | 71 | |
BC138517.1 | - | 1 - 2 | 235 | 71 | |
U31758.1 | - | 1 - 2 | 223 | 71 |