The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162965.1 | NP_001156437.1 | 4 - 5 | 399 | 64 |
NM_001254731.1 | NP_001241660.1 | 4 - 5 | 399 | 64 | |
NM_172718.3 | NP_766306.2 | 4 - 5 | 399 | 64 | |
GenBank mRNA | AK044716.1 | - | 4 - 5 | 353 | 64 |
AK053555.1 | - | 4 - 5 | 333 | 64 | |
AK170021.1 | - | 4 - 5 | 391 | 64 | |
BC138049.1 | - | 4 - 5 | 325 | 64 | |
BC138050.1 | - | 4 - 5 | 325 | 64 |