Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kctd17
Entrez Gene ID
72844
Gene Name
potassium channel tetramerisation domain containing 17
Gene Aliases
2900008M13Rik, AA414907, AW742389, N28155
Chromosome Location
Chr.15: 78428559 - 78439303 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289671.1 NP_001276600.1 3 - 4 509 66
NM_001289672.1 NP_001276601.1 3 - 4 509 66
NM_001289673.1 NP_001276602.1 3 - 4 509 66
NR_110357.1 3 - 4 509 66
GenBank mRNA AK179755.1 - 2 - 3 197 66
BC144818.1 - 3 - 4 509 66
BC144820.1 - 3 - 4 509 66
BC145799.1 - 3 - 4 509 66

View Full Product Details