The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080746.2 | NP_001074215.1 | 25 - 26 | 2620 | 70 |
NM_001080747.2 | NP_001074216.1 | 24 - 25 | 2557 | 70 | |
NM_001080748.2 | NP_001074217.1 | 23 - 24 | 2500 | 70 | |
NM_001080749.2 | NP_001074218.1 | 23 - 24 | 2500 | 70 | |
NM_010365.4 | NP_034495.2 | 24 - 25 | 2563 | 70 | |
GenBank mRNA | AF043219.1 | - | 23 - 24 | 2184 | 70 |
AF043220.1 | - | 22 - 23 | 2121 | 70 | |
AK013348.1 | - | 2 - 3 | 85 | 70 | |
AK145698.1 | - | 15 - 16 | 1448 | 70 | |
AK147201.1 | - | 23 - 24 | 2458 | 70 | |
AK180983.1 | - | 2 - 3 | 139 | 70 | |
AY030290.1 | - | 24 - 25 | 2250 | 70 | |
AY030291.1 | - | 24 - 25 | 2256 | 70 | |
AY030292.1 | - | 23 - 24 | 2193 | 70 | |
AY030293.1 | - | 25 - 26 | 2313 | 70 | |
BC047387.1 | - | 23 - 24 | 2148 | 70 | |
BC053044.1 | - | 25 - 26 | 2589 | 70 |