The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080746.2 | NP_001074215.1 | 5 - 6 | 937 | 71 |
NM_001080747.2 | NP_001074216.1 | 5 - 6 | 937 | 71 | |
NM_001080748.2 | NP_001074217.1 | 5 - 6 | 937 | 71 | |
NM_001080749.2 | NP_001074218.1 | 5 - 6 | 937 | 71 | |
NM_010365.4 | NP_034495.2 | 5 - 6 | 937 | 71 | |
GenBank mRNA | AF017085.1 | - | 6 - 7 | 641 | 71 |
AF043219.1 | - | 4 - 5 | 558 | 71 | |
AF043220.1 | - | 4 - 5 | 558 | 71 | |
AK147201.1 | - | 5 - 6 | 895 | 71 | |
AY030290.1 | - | 5 - 6 | 630 | 71 | |
AY030291.1 | - | 5 - 6 | 630 | 71 | |
AY030292.1 | - | 5 - 6 | 630 | 71 | |
AY030293.1 | - | 5 - 6 | 630 | 71 | |
BC047387.1 | - | 5 - 6 | 585 | 71 | |
BC053044.1 | - | 5 - 6 | 906 | 71 |