Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Hspb1
- Entrez Gene ID
-
15507
- Gene Name
- heat shock protein 1
- Gene Aliases
-
27kDa, Hsp25
- Chromosome Location
-
Chr.5: 135887919 - 135889563 on Build GRCm38
- Gene Symbol
- Gm9817
- Entrez Gene ID
-
100303734
- Gene Name
- predicted gene 9817
- Gene Aliases
-
ENSMUSG00000047061
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK019498.1 |
- |
1 - 1 |
|
67 |
View Full Product Details