Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbx5
Entrez Gene ID
21388
Gene Name
T-box 5
Gene Aliases
-
Chromosome Location
Chr.5: 119796515 - 119885220 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011537.3 NP_035667.1 3 - 4 899 69
GenBank mRNA AF140427.1 - 3 - 4 664 69
BC090639.1 - 3 - 4 903 69

View Full Product Details