The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010324.2 | NP_034454.2 | 6 - 7 | 942 | 60 |
GenBank mRNA | AK146445.1 | - | 6 - 7 | 844 | 60 |
AK147807.1 | - | 6 - 7 | 864 | 60 | |
AK147934.1 | - | 6 - 7 | 847 | 60 | |
AK186873.1 | - | 3 - 4 | 60 | ||
BC002057.1 | - | 6 - 7 | 830 | 60 | |
J02623.1 | - | 6 - 7 | 846 | 60 |