The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175353.2 | NP_780562.2 | 21 - 22 | 2336 | 70 |
GenBank mRNA | AK076455.1 | - | 4 - 5 | 509 | 70 |
AK151524.1 | - | 21 - 22 | 2337 | 70 | |
AK151838.1 | - | 21 - 22 | 2337 | 70 | |
BC025616.1 | - | 8 - 9 | 910 | 70 | |
BC026859.1 | - | 21 - 22 | 2289 | 70 |