The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175353.2 | NP_780562.2 | 12 - 13 | 1259 | 143 |
GenBank mRNA | AK082881.1 | - | 12 - 13 | 1238 | 143 |
AK151524.1 | - | 12 - 13 | 1260 | 143 | |
AK151838.1 | - | 12 - 13 | 1260 | 143 | |
BC026859.1 | - | 12 - 13 | 1212 | 143 |