Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ift27
Entrez Gene ID
67042
Gene Name
intraflagellar transport 27
Gene Aliases
2600013G09Rik, Rabl4
Chromosome Location
Chr.15: 78159463 - 78174108 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025931.2 NP_080207.1 4 - 5 536 68
GenBank mRNA AK011196.1 - 4 - 5 534 68
AK044278.1 - 1 - 2 974 68
BC017514.1 - 4 - 5 536 68

View Full Product Details