Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aff1
Entrez Gene ID
17355
Gene Name
AF4/FMR2 family, member 1
Gene Aliases
9630032B01Rik, AW319193, Af4, Mllt2h, Rob
Chromosome Location
Chr.5: 103692334 - 103855322 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080798.1 NP_001074267.1 6 - 7 1322 108
NM_133919.3 NP_598680.3 6 - 7 1333 108
GenBank mRNA AK143742.1 - 6 - 7 1322 108
AY749168.1 - 6 - 7 1166 108
BC131665.1 - 6 - 7 1166 108
CD543840.1 - 4 - 5 447 108

View Full Product Details