Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sfswap
Entrez Gene ID
231769
Gene Name
splicing factor, suppressor of white-apricot homolog (Drosophila)
Gene Aliases
1190005N23Rik, 6330437E22Rik, AI197402, AW212079, SWAP, Sfrs8, Srsf8
Chromosome Location
Chr.5: 129501231 - 129571384 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172276.3 NP_758480.2 10 - 11 1700 60
GenBank mRNA AK031872.1 - 10 - 11 1691 60
AK088182.1 - 10 - 11 1644 60
AK140369.1 - 10 - 11 1693 60
BC023268.1 - 5 - 6 731 60
BC057314.1 - 10 - 11 3611 60

View Full Product Details