The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005426.2 | NP_001005426.2 | 7 - 8 | 899 | 57 |
GenBank mRNA | BC071186.1 | - | 7 - 8 | 826 | 57 |