Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zcwpw1
Entrez Gene ID
381678
Gene Name
zinc finger, CW type with PWWP domain 1
Gene Aliases
Gm1053
Chromosome Location
Chr.5: 137787395 - 137822621 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005426.2 NP_001005426.2 8 - 9 1012 90
GenBank mRNA BC071186.1 - 8 - 9 939 90

View Full Product Details