Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp2c70
Entrez Gene ID
226105
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 70
Gene Aliases
-
Chromosome Location
Chr.19: 40153361 - 40187286 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145499.2 NP_663474.2 4 - 5 665 148
GenBank mRNA AK050082.1 - 4 - 5 666 148
AY227736.1 - 4 - 5 649 148
BC016494.1 - 4 - 5 650 148
BC022151.1 - 4 - 5 643 148
BC023894.1 - 4 - 5 657 148
BC025822.1 - 4 - 5 667 148
BC034831.1 - 4 - 5 649 148

View Full Product Details