Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp2c70
Entrez Gene ID
226105
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 70
Gene Aliases
-
Chromosome Location
Chr.19: 40153361 - 40187286 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145499.2 NP_663474.2 5 - 6 838 104
GenBank mRNA AK050082.1 - 5 - 6 839 104
AY227736.1 - 5 - 6 822 104
BC016494.1 - 5 - 6 823 104
BC022151.1 - 5 - 6 816 104
BC023894.1 - 5 - 6 830 104
BC025822.1 - 5 - 6 840 104
BC034831.1 - 5 - 6 822 104

View Full Product Details