The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009077.2 | NP_033103.2 | 3 - 4 | 263 | 57 |
GenBank mRNA | AK007263.1 | - | 3 - 4 | 233 | 57 |
AK009044.1 | - | 3 - 4 | 234 | 57 | |
AK010510.1 | - | 3 - 4 | 231 | 57 | |
AK012580.1 | - | 3 - 4 | 231 | 57 | |
AK145649.1 | - | 3 - 4 | 264 | 57 | |
AK151667.1 | - | 3 - 4 | 233 | 57 | |
AK153172.1 | - | 3 - 4 | 231 | 57 | |
AK166803.1 | - | 3 - 4 | 233 | 57 | |
AK168288.1 | - | 3 - 4 | 231 | 57 | |
AK169659.1 | - | 3 - 4 | 231 | 57 | |
BC072558.1 | - | - | 2572 | 57 | |
BC081468.1 | - | 3 - 4 | 221 | 57 | |
BC082290.1 | - | 3 - 4 | 217 | 57 | |
BC091732.1 | - | 2 - 3 | 209 | 57 | |
DQ403025.1 | - | 2 - 3 | 128 | 57 |