Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klk6
Entrez Gene ID
19144
Gene Name
kallikrein related-peptidase 6
Gene Aliases
AI849898, BSP, Bssp, Klk29, MSP, Prss18, Prss9, neurosin
Chromosome Location
Chr.7: 43824544 - 43832027 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164697.1 NP_001158169.1 1 - 2 45 69
GenBank mRNA AK138218.1 - 1 - 2 46 69

View Full Product Details