The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008118.3 | NP_032144.2 | 8 - 9 | 1276 | 84 |
GenBank mRNA | AK078933.1 | - | 8 - 9 | 1277 | 84 |
BC118519.1 | - | 8 - 9 | 1262 | 84 | |
L24191.1 | - | 8 - 9 | 1188 | 84 |