Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ms4a10
Entrez Gene ID
69826
Gene Name
membrane-spanning 4-domains, subfamily A, member 10
Gene Aliases
2010001N17Rik
Chromosome Location
Chr.19: 10962289 - 10974725 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023529.2 NP_076018.2 3 - 4 366 90
GenBank mRNA AF237918.1 - 3 - 4 366 90
BC144935.1 - 3 - 4 363 90
BC150675.1 - 3 - 4 363 90

View Full Product Details