The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133900.4 | NP_598661.1 | 5 - 6 | 905 | 77 |
GenBank mRNA | AK088865.1 | - | 5 - 6 | 857 | 77 |
AK168048.1 | - | 3 - 4 | 513 | 77 | |
BC002251.1 | - | 3 - 4 | 476 | 77 | |
BQ177662.1 | - | 3 - 4 | 471 | 77 |