The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037758.2 | NP_001032847.2 | 8 - 9 | 1081 | 64 |
NM_001286465.1 | NP_001273394.1 | 8 - 9 | 1081 | 64 | |
NM_001286466.1 | NP_001273395.1 | 7 - 8 | 849 | 64 | |
NM_009771.3 | NP_033901.1 | 7 - 8 | 973 | 64 | |
GenBank mRNA | AF081887.1 | - | 7 - 8 | 958 | 64 |
AF099932.1 | - | 7 - 8 | 868 | 64 | |
AF110396.1 | - | 7 - 8 | 868 | 64 | |
AF112979.1 | - | 7 - 8 | 868 | 64 | |
AK032221.1 | - | 8 - 9 | 1044 | 64 | |
AK052317.1 | - | 9 - 10 | 1105 | 64 | |
AK083257.1 | - | 8 - 9 | 1057 | 64 | |
AK145091.1 | - | 7 - 8 | 785 | 64 | |
AK145624.1 | - | 8 - 9 | 1064 | 64 | |
AK156660.1 | - | 7 - 8 | 1001 | 64 | |
AK220183.1 | - | 8 - 9 | 1079 | 64 | |
BC003989.1 | - | 7 - 8 | 874 | 64 |