Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdzd7
Entrez Gene ID
100503041
Gene Name
PDZ domain containing 7
Gene Aliases
9130207N01, EG435601, Pdzk7
Chromosome Location
Chr.19: 45026907 - 45058308 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195265.1 NP_001182194.1 4 - 5 842 103
GenBank mRNA KF041446.1 - 4 - 5 713 103
KF041447.1 - 4 - 5 713 103
KF041448.1 - 3 - 4 572 103
KF041449.1 - 4 - 5 713 103
KF041450.1 - 4 - 5 713 103
KF041451.1 - 4 - 5 713 103

View Full Product Details