The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174870.3 | NP_777359.3 | 1 - 2 | 409 | 67 |
GenBank mRNA | AK085280.1 | - | 1 - 2 | 117 | 67 |
AK128888.1 | - | 1 - 2 | 129 | 67 | |
AK143528.1 | - | 1 - 2 | 137 | 67 | |
AY093420.1 | - | 1 - 2 | 329 | 67 | |
BC022130.1 | - | 1 - 2 | 98 | 67 | |
BC032151.1 | - | 1 - 2 | 137 | 67 |