The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145542.3 | NP_663517.2 | 11 - 12 | 1393 | 60 |
GenBank mRNA | AB092504.1 | - | 11 - 12 | 1122 | 60 |
AK078823.1 | - | 4 - 5 | 595 | 60 | |
BC018218.1 | - | 11 - 12 | 1222 | 60 |