The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033313.3 | NP_001028485.1 | 1 - 2 | 120 | 101 |
GenBank mRNA | AK147170.1 | - | 1 - 2 | 121 | 101 |
BC139204.1 | - | 1 - 2 | 73 | 101 | |
BC139205.1 | - | 1 - 2 | 73 | 101 |