The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010158.2 | NP_034288.2 | 6 - 7 | 1137 | 78 |
GenBank mRNA | AF079763.1 | - | 6 - 7 | 1145 | 78 |
AF099092.1 | - | 6 - 7 | 1148 | 78 | |
AK014353.1 | - | 6 - 7 | 1123 | 78 | |
AK132792.1 | - | 6 - 7 | 839 | 78 | |
AK160564.1 | - | 3 - 4 | 484 | 78 | |
BC031507.1 | - | 6 - 7 | 883 | 78 | |
BC057577.1 | - | 6 - 7 | 1121 | 78 |