Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tgfbr3
Entrez Gene ID
21814
Gene Name
transforming growth factor, beta receptor III
Gene Aliases
1110036H20Rik, AU015626, AW215636, TBRIII
Chromosome Location
Chr.5: 107106570 - 107289595 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011578.3 NP_035708.2 8 - 9 1512 73
GenBank mRNA AF039601.1 - 8 - 9 1312 73
AK133234.1 - 6 - 7 1022 73
BC070428.1 - 8 - 9 1437 73

View Full Product Details