Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Neurl1a
- Entrez Gene ID
-
18011
- Gene Name
- neuralized homolog 1A (Drosophila)
- Gene Aliases
-
2410129E16Rik, AI450910, AI481072, Neu1, Neur1, Neurl, Neurl1, Nlz, Rnf67
- Chromosome Location
-
Chr.19: 47178820 - 47259441 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK010787.1 |
- |
3 - 4 |
962 |
89 |
View Full Product Details