Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aacs
Entrez Gene ID
78894
Gene Name
acetoacetyl-CoA synthetase
Gene Aliases
2210408B16Rik, SUR5
Chromosome Location
Chr.5: 125475873 - 125517403 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030210.1 NP_084486.1 13 - 14 1529 92
GenBank mRNA AB054122.1 - 13 - 14 1487 92
AB056114.1 - 13 - 14 1424 92
AK019063.1 - 13 - 14 1529 92
AK034184.1 - 13 - 14 1462 92
AK043848.1 - 13 - 14 1552 92
AK044017.1 - 13 - 14 1529 92
AK147189.1 - 13 - 14 1529 92
AK150266.1 - 13 - 14 1529 92
AK165418.1 - 13 - 14 1529 92
AK165457.1 - 13 - 14 1529 92
AK170120.1 - 13 - 14 1529 92
AK170653.1 - 13 - 14 1529 92
AK170856.1 - 13 - 14 1529 92
AY091469.1 - 3 - 4 276 92
BC026817.1 - 13 - 14 1520 92

View Full Product Details