Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dao
Entrez Gene ID
13142
Gene Name
D-amino acid oxidase
Gene Aliases
AI987963, DAAO, DAMOX, Dao-1, Dao1
Chromosome Location
Chr.5: 113999636 - 114025675 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286397.1 NP_001273326.1 1 - 2 52 84
NM_010018.3 NP_034148.2 1 - 2 201 84
GenBank mRNA AK134813.1 - 1 - 2 62 84
BB843516.1 - 1 - 2 96 84
BC015269.1 - 1 - 2 52 84
BE651085.1 - 1 - 2 192 84
M32299.1 - 1 - 2 58 84

View Full Product Details