The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028386.1 | NP_082662.1 | 1 - 2 | 1057 | 71 |
GenBank mRNA | AK013495.1 | - | 1 - 2 | 309 | 71 |
BC046606.1 | - | 1 - 2 | 1057 | 71 |