Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Caln1
Entrez Gene ID
140904
Gene Name
calneuron 1
Gene Aliases
9630012C17Rik, Cabp8, MNCb-0849
Chromosome Location
Chr.5: 130369356 - 130846325 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK081011.1 - 1 - 2 102 62

View Full Product Details