Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam175a
Entrez Gene ID
70681
Gene Name
family with sequence similarity 175, member A
Gene Aliases
3830405G04Rik, 5630400M01Rik, AI506069, AL024423, AV118690, Ccdc98
Chromosome Location
Chr.5: 100804802 - 100845216 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172405.3 NP_765993.1 2 - 3 196 119
GenBank mRNA AK014420.1 - 2 - 3 222 119
AK042001.1 - 2 - 3 205 119
AK042339.1 - 2 - 3 205 119
AK051816.1 - 2 - 3 197 119
AK167540.1 - 2 - 3 227 119
BC029845.1 - 2 - 3 201 119

View Full Product Details