Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam175a
Entrez Gene ID
70681
Gene Name
family with sequence similarity 175, member A
Gene Aliases
3830405G04Rik, 5630400M01Rik, AI506069, AL024423, AV118690, Ccdc98
Chromosome Location
Chr.5: 100804802 - 100845216 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172405.3 NP_765993.1 7 - 8 698 138
GenBank mRNA AK051816.1 - 7 - 8 699 138
AK167540.1 - 7 - 8 729 138
BC029845.1 - 7 - 8 703 138

View Full Product Details